
N-Isopropylhydroxylamine hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50632-53-6
:
Formula:C₃H₉NO
MDL Number:MFCD00012599
:111.57 g/mol
Melting Pt:68…72 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige Adhering crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 %
Specification Test Results
Frequently Bought Together