1,5-Bis(methylamino)-3-oxapentane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2620-27-1
:
Formula:C₆H₁₆N₂O
MDL Number:MFCD00142559
:132.207 g/mol
Boiling Pt:76…78 °C (20 mmHg)
Density:0,872 g/cm³
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4370 to 1.4390 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together