
N-(tert-Butoxycarbonyl)-L-phenylalanine 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13734-34-4
:
Formula:C₁₄H₁₉NO₄
MDL Number:MFCD00002663
:265.31 g/mol
Melting Pt:85…88 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:85 to 88 °C
- Water:≤0.5 % (K.F.)
- HPLC:≥99.0 %
- Specific optical rotation:+24° to +26° (20 °C, 589 nm) (c=1, ethanol)
- Titration with NaOH:≥99.0 %
Specification Test Results
Frequently Bought Together