ContactUsLinkComponent
Iodine solution 0,1 mol ICl/l acc. Wijs
undefined
Iodine solution 0,1 mol ICl/l acc. Wijs
  • Appearance:
    Clear red-brown to brown Solution
  • Titration with Na2S2O3:
    0.098 to 0.113 M
Frequently Bought Together