Ethyl-cis-2-amino-1-cyclohexanecarboxylate hydrochloride 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1127-99-7
:
Formula:C₉H₁₈ClNO₂
MDL Number:MFCD00144280
:207.698 g/mol
Melting Pt:128…135 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:128 to 135 °C
- Titration Argentometric:≥94 %
Specification Test Results
Frequently Bought Together