
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:39637-99-5
:
Formula:C₁₀H₈ClF₃O₂
MDL Number:MFCD00044400
:252.62 g/mol
Boiling Pt:213…214 °C (760 mmHg)
Density:1,34 g/cm³
Flash Pt:89 °C
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4675 to 1.4715 (20°C, 589 nm)
- GC:≥98.0 %
- Specific optical rotation:-128° to -134° (20°C, 589 nm) (c=1, CHCl3)
Specification Test Results
Frequently Bought Together