
Isonicotinic anhydride 90% remainder picolinic and nicotinic acid-anhydride
Catalog # ACRO250280050
Supplier: Thermo Fisher Scientific
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Off-white to beige to light brown Crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥88 %
Specification Test Results
About this item
CAS Number:7082-71-5
:
Formula:C₁₂H₈N₂O₃
MDL Number:MFCD00062784
:228.207 g/mol
Melting Pt:133 °C
:GHS05
Storage Temperature:Ambient