
(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4272-74-6
:
Formula:C₁₄H₂₂Cl₂N₂O₃S
MDL Number:MFCD00065395
:369.301 g/mol
Melting Pt:160…161 °C (dec.)
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to light beige Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 %
- Appearance of solution:(5% soln. in methanol) clear, slightly colored
- Specific optical rotation:-9° to -10° (24°C, 589 nm) (c=5, H2O)
Specification Test Results
Frequently Bought Together