
N-(p-Toluenesulphonyl)-L-phenylalanylchloromethylketone 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:402-71-1
:
Formula:C₁₇H₁₈ClNO₃S
MDL Number:MFCD00000935
:351.85 g/mol
Melting Pt:105…109 °C
:GHS05
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:105 to 109 °C
- TLC:≥98.0 %
- Specific optical rotation:-86° to -88° (20 °C, 589 nm) (c=1, abs. ethanol)
Specification Test Results
Frequently Bought Together