
Puromycin dihydrochloride hydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:58-58-2
:
Formula:C₂₂H₂₉N₇O₅·2HCl·nH₂O
MDL Number:MFCD00150080
:544.44 g/mol (anhy)
Melting Pt:168…178 °C
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to light beige Powder
- Infrared spectrum:Conforms
- Water:≤12 %
- HPLC:≥98.5 %
Specification Test Results
Frequently Bought Together