- Antibody type:Secondary
- Antigen name:Rabbit IgG
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:CF® 405S
- Host:Donkey
- Reactivity:Rabbit
- Cross adsorption:Bovine, chicken, goat, guinea pig, horse, human, mouse, sheep, Syrian hamster
- Form:Lyophilized
- Storage buffer:PBS, 15 mg/ml BSA, 20 mg/mL trehalose (after reconstitution)
- Storage temperature:Store at –20 °C.
- Concentration:2 mg/ml (after reconstitution)
- Purification:Affinity purified
- Size:1 mg
- Pk:1 mg
This product is prepared by labeling highly cross adsorbed donkey anti-rabbit IgG (H L) with CF™405S. CF™405S is a blue fluorescent dye with an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition, the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result, flow cytometry analysis using CF™405S dye results in much brighter signal than that using Alexa Fluor® 405. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, human, mouse, rat, and sheep serum proteins.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF405S
Epitope:
Host: Donkey
Isotype:
Reactivity: Rabbit