Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-IgG Llama Polyclonal Antibody (CF™ 594)
Anti-IgG Llama Polyclonal Antibody (CF™ 594)
Supplier:  Biotium
Certificates
Anti-IgG Llama Polyclonal Antibody (CF™ 594)
Supplier:  Biotium

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • 20456-50UL
  • 20456-500UL
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Mouse IgG
  • Mouse IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Llama
  • Llama
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Affinity purified
  • Affinity purified
  • Concentration:
  • 2 mg/mL
  • 2 mg/mL
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • Storage temperature:
  • Store at –20 °C.
  • Store at –20 °C.

Specifications

Frequently Bought Together