Anti-Intestinal FABP Antibody
Catalog # BSBTPB9943
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 2, Intestinal
- Antigen symbol:FABP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2169
- Antigen synonyms:I-FABP|FABPI
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Fatty acid-binding protein, intestinal(FABP2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: FABP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat