ContactUsLinkComponent
Western blot roller, Pierce™
Western blot roller, Pierce™
Catalog # PIER84747
undefined
Western blot roller, Pierce™
Catalog # PIER84747
Specifications
  • Description:
    Western blot roller, PTFE, 8,4 mm wide, with stainless steel handle
  • Pk:
    1
Specifications
About this item
The Western blot roller is a simple, reusable tool for gently and easily removing trapped air bubbles when assembling transfer stacks in semi-dry or wet-tank transfer cassettes.
  • Reusable
  • Durable
The transfer-stack roller has a stainless steel handle and an inert PTFE roller that is 8,4 cm wide. Mini-gel sandwiches can be rolled out with one or two passes, but the tool also works well for transfer stacks with larger gels.
Caution: For Research Use Only. Not for use in diagnostic procedures.