Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt), Pierce™
:106627-54-7
:
:C₄H₄NNaO₆S
:MFCD00043100
:217.13 g/mol
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:Carbodiimide
- Spacer:Short
- Form:Powder
- Quantity:500 mg
- Type:Crosslinker
- Solubility:Water
Frequently Bought Together