Orders LinkContactUsLinkComponent
Block mailer, paraffin
Block mailer, paraffin
Block mailer for 6 paraffin blocks, 134×141×29 mm
Catalog # SIMPM477-6
Supplier:   Simport Scientific
Block mailer, paraffin
Catalog # SIMPM477-6
Supplier:  Simport Scientific
Block mailer, paraffin
Block mailer, paraffin
Block mailer for 6 paraffin blocks, 134×141×29 mm
Price
£141.00
Log in for availability
1 * 50 items
Product Details & Documents
PVC, translucent.
  • Sturdy, secure and easy to use
  • Easy viewing of contents
This mailer can be used for handling and shipping up to 6 blocks as needed and is suitable for all regular models of tissue and biopsy cassettes.
Documents
Specifications
  • Description:
    Block mailer for 6 paraffin blocks
  • W×D×H:
    134×141×29 mm