L-Alanine USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:56-41-7
:
Formula:C₃H₇NO₂
MDL Number:MFCD00064410
:89.09 g/mol
Merck Index:13,00203
Boiling Pt:189 °C (1013 hPa)
Melting Pt:79 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-273-8
Specification Test Results
- Assay:98.5% Min.
Specification Test Results
Frequently Bought Together