Polyethylene glycol 8,000 NF
: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000
:25322-68-3
:MFCD00081839
:H(OCH₂CH₂)nOH
:MFCD01779615
:250
:1.125 g/cm³ (20 °C)
:8000 g/mol
:12,07729
:Ambient
:500-038-2
- Environmentally Preferable:
Frequently Bought Together