Kerosene low odor
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: C13-14 isoparaffin, Light distillate
CAS Number:64742-47-8
:
Merck Index:14,5294
Boiling Pt:200…250 °C
Density:0,800
Flash Pt:76 °C (169 °F)
Storage Temperature:Ambient
EINECS:265-149-8
UN:1223
ADR:3,III
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together