
6-Methoxy-2-naphthylglyoxal hydrate 98% (dry weight)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:745783-88-4
:
Formula:C₁₃H₁₂O₄
MDL Number:MFCD05664091
:232.24 g/mol
Melting Pt:125…130 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together