
(1S,4S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:132747-20-7
:
Formula:C₅H₁₀N₂
MDL Number:MFCD01321289
Melting Pt:∼ 290 °C (dec.)
:259.97 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together