Order Entry
Northern Ireland
ContactUsLinkComponent
1-Bromo-4-(tert-butoxy)benzene 98%
Irritant
1-Bromo-4-(tert-butoxy)benzene 98%
Catalog # L20158.06
CAS Number:  
Irritant
1-Bromo-4-(tert-butoxy)benzene 98%
Catalog # L20158.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :229.12 g/mol
Boiling Pt:63…65 °C (0,4 mmHg)
 :GHS07
Storage Temperature:Ambient
CAS Number:60876-70-2
Formula:C₁₀H₁₃BrO
MDL Number:MFCD00792676