

(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate 97%
Catalog # L20060.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:247.36 g/mol
Melting Pt:33…36 °C
Density:1,106
ADR:8,III
:GHS05
:GHS07
CAS Number:737000-89-4
Formula:C₁₄H₁₇NOS
MDL Number:MFCD05664047