
2-(4-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:138500-85-3
:
Formula:C₁₃H₁₈BBrO₂
MDL Number:MFCD02179493
:297.00 g/mol
Melting Pt:83…85 °C
Storage Temperature:Ambient
ADR:8,III
:GHS05
Related Information
Frequently Bought Together