Bis(phenylacetyl)disulphide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15088-78-5
:
Formula:C₁₆H₁₄O₂S₂
MDL Number:MFCD00513572
:302.41 g/mol
Melting Pt:58…60 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together