Order Entry
Northern Ireland
ContactUsLinkComponent
(S)-1-Methyl-3-pyrrolidinol 98%, ee 99%
Irritant
(S)-1-Methyl-3-pyrrolidinol 98%, ee 99%
Catalog # L19695.MD
CAS Number:  
Irritant
(S)-1-Methyl-3-pyrrolidinol 98%, ee 99%
Catalog # L19695.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

CAS Number:104641-59-0
 :
Formula:C₅H₁₁NO
MDL Number:MFCD03788748
 :101.15 g/mol
Boiling Pt:103…106 °C (35 mmHg)
Density:0,920
Flash Pt:70 °C (158 °F)
 :GHS07
Storage Temperature:Ambient