
(R)-1-Methyl-3-pyrrolidinol ≥99%, ee 99%
Catalog # L19694.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:101.15 g/mol
Boiling Pt:103…106 °C (35 mmHg)
Density:0,920
Flash Pt:70 °C (158 °F)
EINECS:000-000-0
:GHS07
Storage Temperature:Ambient
CAS Number:104641-60-3
Formula:C₅H₁₁NO
MDL Number:MFCD03788747