tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:330793-01-6
:
Formula:C₁₇H₂₆BNO₄
MDL Number:MFCD02179439
:319.208 g/mol
Melting Pt:169…170 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together