L-Proline-tert-butyl ester hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5497-76-7
:
Formula:C₉H₁₇NO₂
MDL Number:MFCD00153459
:207.70 g/mol
Melting Pt:110…112 °C
Storage Temperature:Ambient
EINECS:226-835-2
Related Information
Frequently Bought Together