N-Boc-L-leucinol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:82010-31-9
:
Formula:C₁₁H₂₃NO₃
MDL Number:MFCD00076950
:217.3 g/mol
Boiling Pt:212…214 °C
Density:0,980
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
Related Information
Frequently Bought Together