


4-(Trifluoromethylthio)phenyl isocyanate 97%
Catalog # L19206.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:24032-84-6
:
Formula:C₈H₄F₃NOS
MDL Number:MFCD00036025
:219.19 g/mol
Boiling Pt:175 °C
Density:1,385
Flash Pt:80 °C (176 °F)
ADR:6.1,III
:GHS06
:GHS08
:GHS07