
Harmine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:442-51-3
:
Formula:C₁₃H₁₂N₂O
MDL Number:MFCD00004958
:212.25 g/mol
Merck Index:14,4616
Melting Pt:262…266 °C
EINECS:207-131-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together