2-Allyl-4,6-dibenzoylresorcinol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:102593-74-8
:
Formula:C₂₃H₁₈O₄
MDL Number:MFCD02094038
:358.40 g/mol
Melting Pt:158…163 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together