
Bis(tetramethylene)fluoroformamidinium hexafluorophosphate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:164298-25-3
:
Formula:C₉H₁₆F₇N₂P
MDL Number:MFCD02683430
:316.20 g/mol
Melting Pt:∼ 155 °C
ADR:8,III
:GHS05
Related Information
Frequently Bought Together