
5-Ethoxy-2-mercaptobenzimidazole 97%
Catalog # L17781.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:55489-15-1
:
Formula:C₉H₁₀N₂OS
MDL Number:MFCD01326487
:194.25 g/mol
Melting Pt:246…250 °C
:GHS07
Storage Temperature:Ambient