Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dihydroxybenzaldoxime
3,4-Dihydroxybenzaldoxime 98%
3,4-Dihydroxybenzaldoxime 98%
3,4-Dihydroxybenzaldoxime 98%
L17766.06
:
Thermo Fisher Scientific
:
3,4-Dihydroxybenzaldoxime 98%
L17766.06
:
Thermo Fisher Scientific
:
Pk:
5 g
Seq:
3173
:
3343-59-7
:
:
C₇H₇NO₃
:
MFCD01070622
:
153.14 g/mol
:
155…157 °C (dec.)
:
GHS07
:
Ambient