Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Boc-indoline
1-Boc-indoline 98%
1-Boc-indoline 98%
1-Boc-indoline 98%
L17477.06
:
Thermo Fisher Scientific
:
1-Boc-indoline 98%
L17477.06
:
Thermo Fisher Scientific
:
Pk:
5 g
Seq:
3173
:
:
219.29 g/mol
:
150 °C (15 mmHg)
:
45…49 °C
:
>110 °C (230 °F)
:
GHS07
:
143262-10-6
:
C₁₃H₁₇NO₂
:
MFCD01318399