
4-Bromo-2-fluorophenylboronic acid ≥98%
Catalog # L17468.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:216393-64-5
:
Formula:C₆H₅BBrFO₂
MDL Number:MFCD01318110
:218.82 g/mol
Melting Pt:238…242 °C
:GHS07
Storage Temperature:Ambient