
D-Proline methyl ester hydrochloride 99%
Catalog # L17259.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:65365-28-8
:
Formula:C₆H₁₁NO₂
MDL Number:MFCD00083685
:165.62 g/mol
Melting Pt:69…71 °C
:GHS07