Order Entry
Northern Ireland
ContactUsLinkComponent
(1S,2S)-(+)-2-Benzyloxycyclopentylamine ≥99%, ee 99%, ChiPros®
Corrosive
(1S,2S)-(+)-2-Benzyloxycyclopentylamine ≥99%, ee 99%, ChiPros®
Catalog # L17018.03
CAS Number:  
Corrosive
(1S,2S)-(+)-2-Benzyloxycyclopentylamine ≥99%, ee 99%, ChiPros®
Catalog # L17018.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:181657-57-8
 :
Formula:C₁₂H₁₇NO
MDL Number:MFCD01075751
 :191.27 g/mol
Boiling Pt:90…93 °C (0,23 mmHg)
Melting Pt:23 °C
Density:0,983
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:2735
ADR:8,III
 :GHS05