

Antimony(V) fluoride compound with graphite (1:1)
Catalog # L16962.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
CAS Number:59839-60-0
:
Formula:C:SbF₅
MDL Number:MFCD00236643
:228.75 g/mol
ADR:8,II
:GHS06
:GHS05
Storage Temperature:Ambient