
Lithium trifluoroacetate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2923-17-3
:
Formula:LiCO₂CF₃
MDL Number:MFCD00013216
:119.96 g/mol
Storage Temperature:Ambient
EINECS:220-878-0
UN:2811
ADR:6.1,II
:GHS06
Related Information
Frequently Bought Together