
Methyl pentafluorophenyl carbonate 97%
Catalog # L16107.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:36919-03-6
:
Formula:C₈H₃F₅O₃
MDL Number:MFCD01075723
:242.10 g/mol
Boiling Pt:84 °C (3 mmHg)
Melting Pt:27…31 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07