



trans-1,4-Cyclohexane diisocyanate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7517-76-2
:
Formula:C₈H₁₀N₂O₂
MDL Number:MFCD00074787
Boiling Pt:145…146 °C (32 mmHg)
Melting Pt:59…62 °C
Storage Temperature:Ambient
ADR:6.1,II
:166.18 g/mol
:GHS06
:GHS08
:GHS05
:GHS07
Related Information
Frequently Bought Together