£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
2,2,7-Trimethyloctane-3,5-dione
2,2,7-Trimethyloctane-3,5-dione ≥98%
2,2,7-Trimethyloctane-3,5-dione ≥98%
L14639.06
:
Thermo Fisher Scientific
:
2,2,7-Trimethyloctane-3,5-dione ≥98%
L14639.06
:
Thermo Fisher Scientific
:
Pk:
5 g
Seq:
3173
:
69725-37-7
:
:
C₁₁H₂₀O₂
:
MFCD00511279
:
184.28 g/mol
:
44 °C (0,2 mmHg)
:
Ambient