4,5-Dimethoxy-2-nitrobenzyl carbonochloridate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:42855-00-5
:
Formula:C₁₀H₁₀ClNO₆
MDL Number:MFCD00143507
Melting Pt:∼ 120 °C (dec.)
ADR:8,II
:275.65 g/mol
:GHS05
Related Information
Frequently Bought Together