Methyl-4-Chloro-3-methylbenzoate 97%
Catalog # L13110.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:184.62 g/mol
Storage Temperature:Ambient
CAS Number:91367-05-4
Formula:C₉H₉ClO₂
MDL Number:MFCD00151838