Order Entry
Northern Ireland
ContactUsLinkComponent
3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane 97%
3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane 97%
Catalog # L12958.22
CAS Number:  
3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane 97%
Catalog # L12958.22
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

CAS Number:1455-42-1
 :
Formula:C₁₅H₂₈O₆
MDL Number:MFCD00059794
Melting Pt:197…200 °C
 :304.38 g/mol
Storage Temperature:Ambient