

Lithium acetylide ethylenediamine complex 90%, tech.
Catalog # L12401.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:92.07 g/mol
Melting Pt:∼ 76 °C (dec.)
Density:1,04
Storage Temperature:Ambient
EINECS:229-967-9
UN:3131
ADR:4.3,I
:GHS02
:GHS05
CAS Number:6867-30-7
Formula:C₄H₉LiN₂
MDL Number:MFCD00013183