
1H,1H,5H-Octafluoropentyl methacrylate 98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:355-93-1
:
Formula:C₉H₈F₈O₂
MDL Number:MFCD00039278
:300.15 g/mol
Boiling Pt:178…179 °C
Density:1,432
Flash Pt:81 °C (177 °F)
EINECS:206-596-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together